Thermo Scientific Chemicals

2-Hydroxypyridine, 97%, Thermo Scientific Chemicals

Catalog number: 121840500
50 g, Plastic bottle, Each
Thermo Scientific Chemicals

2-Hydroxypyridine, 97%, Thermo Scientific Chemicals

Catalog number: 121840500
50 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 121840500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
142-08-5
Appearance (Form)
Crystalline powder or crystals
Melting point
103°C to 108°C
Appearance (Color)
Off-white to yellow to beige to brown
Titration with HClO4
>=96.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text