Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

PureLink™ DNase Set

Catalog number: 12185010
Have Questions?
Invitrogen™

PureLink™ DNase Set

Catalog number: 12185010
Have Questions?
Catalog Number
12185010
Unit Size
50 Preps
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1218501050 Preps
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The PureLink™ DNase Set provides rapid and efficient removal of DNA from RNA that has been purified using PureLink™ RNA kits. The PureLink™ DNase Set is:

• RNase-free for use in RNA purification
• Optimized for on-column digestion
• Lyophilized for stability

Using the PureLink™ DNase Set
The RNase-free DNase set comes lyophilized and is optimized for on-column digestion of DNA using PureLink™ protocols. The degree of DNA digestion performed by PureLink™ DNase is optional for most PureLink™ RNA kits, due to the efficient removal of the majority of DNA by the PureLink™ technology; however, some downstream applications that are sensitive to small amounts of DNA may require further removal of DNA.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
10X
Shipping Condition
Room Temperature

Contents & Storage

Contains:
• PureLink™ DNase (1500 U, lyophilized)
• 10X DNase I Reaction Buffer (1 mL)
• RNase-Free Water (1 mL)

Store at 4°C upon receipt. After reconstituted, PureLink™ DNase should be stored at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text