Thermo Scientific Chemicals

2-Hydroxypyrimidine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 121861000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Hydroxypyrimidine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 121861000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 121861000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
38353-09-2
IUPAC Name
hydrogen 1,2-dihydropyrimidin-2-one chloride
Molecular Formula
C4H5ClN2O
InChI Key
IAJINJSFYTZPEJ-UHFFFAOYSA-N
SMILES
[H+].[Cl-].O=C1NC=CC=N1
Infrared spectrum
Conforms
Appearance (Color)
Light yellow to beige
Titration with HClO4
>=97.5 %
Appearance (Form)
Crystalline solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text