8-Hydroxyquinoline-5-sulfonic acid monohydrate, 98%, Thermo Scientific Chemicals
8-Hydroxyquinoline-5-sulfonic acid monohydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

8-Hydroxyquinoline-5-sulfonic acid monohydrate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 121921000
Price (USD)/ Each
38.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
45.30 
Save 6.65 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
38.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
45.30 
Save 6.65 (15%)
Add to cart
8-Hydroxyquinoline-5-sulfonic acid monohydrate, 98%, Thermo Scientific Chemicals
Catalog number121921000
Price (USD)/ Each
38.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
45.30 
Save 6.65 (15%)
-
Add to cart
Chemical Identifiers
CAS283158-18-9
IUPAC Name8-hydroxyquinoline-5-sulfonic acid hydrate
Molecular FormulaC9H9NO5S
InChI KeyWUXYGIQVWKDVTJ-UHFFFAOYSA-N
SMILESO.OC1=CC=C(C2=CC=CN=C12)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Infrared spectrumConforms
Assay>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only