3-Hydroxytetrahydrofuran, 97%, Thermo Scientific Chemicals
3-Hydroxytetrahydrofuran, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Hydroxytetrahydrofuran, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 121940050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS453-20-3
IUPAC Nameoxolan-3-ol
Molecular FormulaC4H8O2
InChI KeyXDPCNPCKDGQBAN-UHFFFAOYNA-N
SMILESOC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4490 to 1.4510 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only