Thermo Scientific Chemicals

3-Hydroxytetrahydrofuran, 97%, Thermo Scientific Chemicals

Catalog number: 121941000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Hydroxytetrahydrofuran, 97%, Thermo Scientific Chemicals

Catalog number: 121941000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 121941000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
453-20-3
IUPAC Name
oxolan-3-ol
Molecular Formula
C4H8O2
InChI Key
XDPCNPCKDGQBAN-UHFFFAOYNA-N
SMILES
OC1CCOC1
Infrared spectrum
Conforms
Refractive index
1.4490 to 1.4510 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow
GC
>=96.0 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text