Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This product has been discontinued and is no longer available for purchase.
The Renvo™ SARS-CoV-2 Test is performed on environmental air samples from the Thermo Scientific™ AerosolSense™ Sampler. The test's patented PCR technology enables rapid exponential amplification while reducing overall thermocycling times.
Intended use: For environmental surveillance use only and not for diagnostic purposes.
Contents:
**Not included: Reusable dock, power cord, stand
Control Sets | High/low/neg |
Detectable Analytes | SARS-CoV-2 RNA |
Format | Test Cassette Kit |
Sample Type | Air sample from AerosolSense sample cartridge |
PCR Method | RT-PCR |
Shelf Life | 18 months from date of manufacture |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
121995-01 | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|