3-Hydroxytyramine Hydrochloride, 99%, Thermo Scientific Chemicals
3-Hydroxytyramine Hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Hydroxytyramine Hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
100 g
Packaging:
Glass bottle
Catalog number 122000250
Price (USD)/ Each
95.65
Online exclusive
106.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
95.65
Online exclusive
106.00 
Save 10.35 (10%)
Add to cart
3-Hydroxytyramine Hydrochloride, 99%, Thermo Scientific Chemicals
Catalog number122000250
Price (USD)/ Each
95.65
Online exclusive
106.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS62-31-7
IUPAC Namehydrogen 4-(2-aminoethyl)benzene-1,2-diol chloride
Molecular FormulaC8H12ClNO2
InChI KeyCTENFNNZBMHDDG-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCCC1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=98.5 % (On dried substance)
Loss on drying=<0.5 % (105°C, 2 hrs)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only