Thermo Scientific Chemicals

Hypoxanthine, 99.5%, Thermo Scientific Chemicals

Catalog number: 122011000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Hypoxanthine, 99.5%, Thermo Scientific Chemicals

Catalog number: 122011000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 122011000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68-94-0
IUPAC Name
6,7-dihydro-3H-purin-6-one
Molecular Formula
C5H4N4O
InChI Key
FDGQSTZJBFJUBT-UHFFFAOYSA-N
SMILES
O=C1N=CNC2=C1NC=N2
Appearance (Form)
Crystalline powder
Loss on drying
=<0.5 %
HPLC
>=99.4 %
Sulfated ash
=<0.2 %
Appearance (Color)
White to light yellow to light beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text