Imidazole, 99%, Thermo Scientific Chemicals
Imidazole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Imidazole, 99%, Thermo Scientific Chemicals

Imidazole, >-98.5%, C3H4N2, CAS Number-288-32-4, 1,3-diaza-2,4-cyclopentadiene, 1,3-diazole, glyoxalin, glyoxaline, imidazol, imidazole, iminazole, imutex, miazole, pyrro b monazole, 500g, 99%, 23, 45, 255.0 deg.C to 256.0 deg.C, CHEBI:16069, White to Yellow, 01,492; 02,220, 145 deg.C, 68.08
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
5 g
100 g
500 g
2 kg
Packaging:
View all
Plastic drum
Catalog number 122020100
Price (USD)/ Each
1,229.65
Online exclusive
1366.00 
Save 136.35 (10%)
-
Add to cart
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
1,229.65
Online exclusive
1366.00 
Save 136.35 (10%)
Add to cart
Imidazole, 99%, Thermo Scientific Chemicals
Catalog number122020100
Price (USD)/ Each
1,229.65
Online exclusive
1366.00 
Save 136.35 (10%)
-
Add to cart
Chemical Identifiers
CAS288-32-4
IUPAC Name1H-imidazole
Molecular FormulaC3H4N2
InChI KeyRAXXELZNTBOGNW-UHFFFAOYSA-N
SMILESN1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
GC>=98.5 %
Infrared spectrumConforms
Water=<0.2 % (K.F.)
Appearance (Form)Flakes
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only