1-Indanone, 99+%, Thermo Scientific Chemicals
1-Indanone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Indanone, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Catalog number 122110100
Price (USD)/ Each
54.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
63.80 
Save 9.15 (14%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
54.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
63.80 
Save 9.15 (14%)
Add to cart
1-Indanone, 99+%, Thermo Scientific Chemicals
Catalog number122110100
Price (USD)/ Each
54.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
63.80 
Save 9.15 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Name2,3-dihydro-1H-inden-1-one
Molecular FormulaC9H8O
InChI KeyQNXSIUBBGPHDDE-UHFFFAOYSA-N
SMILESO=C1CCC2=CC=CC=C12
Molecular Weight (g/mol)132.16
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to brown
Appearance (Form)Crystalline mass
GC>=99.0 %
Infrared spectrumConforms
Melting point38°C to 42°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only