Thermo Scientific Chemicals

Indazole, 96%, Thermo Scientific Chemicals

Catalog number: 122120050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Indazole, 96%, Thermo Scientific Chemicals

Catalog number: 122120050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 122120050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
271-44-3
IUPAC Name
1H-indazole
Molecular Formula
C7H6N2
InChI Key
BAXOFTOLAUCFNW-UHFFFAOYSA-N
SMILES
N1N=CC2=CC=CC=C12
Appearance (Color)
White to beige
Titration with HClO4
>=95 %
Melting point
146°C to 150°C
Appearance (Form)
Crystalline powder or needles
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text