Thermo Scientific Chemicals

Indole, 99+%, Thermo Scientific Chemicals

Catalog number: 122150250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Indole, 99+%, Thermo Scientific Chemicals

Catalog number: 122150250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 122150250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
120-72-9
IUPAC Name
1H-indole
Molecular Formula
C8H7N
InChI Key
SIKJAQJRHWYJAI-UHFFFAOYSA-N
SMILES
N1C=CC2=CC=CC=C12
Appearance (Form)
Crystalline powder
Melting point
51°C to 53°C
Appearance (Color)
White to light pink
Infrared spectrum
Conforms
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text