Indole-5-carboxylic acid, 98%, Thermo Scientific Chemicals
Indole-5-carboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indole-5-carboxylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 122210010
Price (USD)/ Each
41.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
48.30 
Save 6.65 (14%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
41.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
48.30 
Save 6.65 (14%)
Add to cart
Indole-5-carboxylic acid, 98%, Thermo Scientific Chemicals
Catalog number122210010
Price (USD)/ Each
41.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
48.30 
Save 6.65 (14%)
-
Add to cart
Chemical Identifiers
CAS1670-81-1
IUPAC Name1H-indole-5-carboxylate
Molecular FormulaC9H6NO2
InChI KeyIENZCGNHSIMFJE-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC=C2NC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration with NaOH>=97.5 %
Melting point207°C to 212°C
Appearance (Color)Yellow to light beige
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only