Indoline, 98%, Thermo Scientific Chemicals
Indoline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indoline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 122245000
Price (USD)/ Each
328.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
387.00 
Save 58.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
328.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
387.00 
Save 58.35 (15%)
Add to cart
Indoline, 98%, Thermo Scientific Chemicals
Catalog number122245000
Price (USD)/ Each
328.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
387.00 
Save 58.35 (15%)
-
Add to cart
Chemical Identifiers
CAS496-15-1
IUPAC Name2,3-dihydro-1H-indole
Molecular FormulaC8H9N
InChI KeyLPAGFVYQRIESJQ-UHFFFAOYSA-N
SMILESC1CC2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5910 to 1.5950 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to brown
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only