Thermo Scientific Chemicals

Inositol, 98+%

Catalog number: 122261000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Inositol, 98+%

Catalog number: 122261000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 122261000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
87-89-8
IUPAC Name
cyclohexane-1,2,3,4,5,6-hexol
Molecular Formula
C6H12O6
InChI Key
CDAISMWEOUEBRE-UHFFFAOYSA-N
SMILES
OC1C(O)C(O)C(O)C(O)C1O
Appearance (Form)
Crystalline powder
Melting point
220°C to 228°C
Loss on drying
=<0.5 %
Solubility
(5 % in water) Clear colorless
HPLC
>=98.0 % (ELSD)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text