Inositol, 98+%
Thermo Scientific Chemicals

Inositol, 98+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 122265000
Price (USD)/ Each
249.65
Online exclusive
277.00 
Save 27.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
249.65
Online exclusive
277.00 
Save 27.35 (10%)
Add to cart
Inositol, 98+%
Catalog number122265000
Price (USD)/ Each
249.65
Online exclusive
277.00 
Save 27.35 (10%)
-
Add to cart
Chemical Identifiers
CAS87-89-8
IUPAC Namecyclohexane-1,2,3,4,5,6-hexol
Molecular FormulaC6H12O6
InChI KeyCDAISMWEOUEBRE-UHFFFAOYSA-N
SMILESOC1C(O)C(O)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point220°C to 228°C
Loss on drying=<0.5 %
Solubility(5 % in water) Clear colorless
HPLC>=98.0 % (ELSD)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only