Thermo Scientific Chemicals

Iodoacetamide, 98%, Thermo Scientific Chemicals

Catalog number: 122270250
25 g, Each
Thermo Scientific Chemicals

Iodoacetamide, 98%, Thermo Scientific Chemicals

Catalog number: 122270250
25 g, Each
Quantity
Catalog number: 122270250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
144-48-9
IUPAC Name
2-iodoacetamide
Molecular Formula
C2H4INO
InChI Key
PGLTVOMIXTUURA-UHFFFAOYSA-N
SMILES
NC(=O)CI
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Melting point
>=90°C
Titration Argentometric
>=97.5 % (ex I)
Appearance (Color)
White to yellow to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text