Iodoacetamide, 98%, Thermo Scientific Chemicals
Iodoacetamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iodoacetamide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
500 g
Catalog number 122271000
Price (USD)/ Each
555.65
Online exclusive
654.00 
Save 98.35 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
555.65
Online exclusive
654.00 
Save 98.35 (15%)
Add to cart
Iodoacetamide, 98%, Thermo Scientific Chemicals
Catalog number122271000
Price (USD)/ Each
555.65
Online exclusive
654.00 
Save 98.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-iodoacetamide
Molecular FormulaC2H4INO
InChI KeyPGLTVOMIXTUURA-UHFFFAOYSA-N
SMILESNC(=O)CI
Molecular Weight (g/mol)184.96
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point>=90°C
Titration Argentometric>=97.5 % (ex I)
Appearance (Color)White to yellow to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only