Thermo Scientific Chemicals

Iodoacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: 122280250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Iodoacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: 122280250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 122280250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64-69-7
IUPAC Name
2-iodoacetic acid
Molecular Formula
C2H3IO2
InChI Key
JDNTWHVOXJZDSN-UHFFFAOYSA-N
SMILES
OC(=O)CI
Appearance (Color)
Yellow to orange-brown
Appearance (Form)
Powder or crystals or flakes
Melting point
76°C to 83°C
Infrared spectrum
Conforms
Titration with NaOH
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text