Iodoacetic acid, 97%, Thermo Scientific Chemicals
Iodoacetic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iodoacetic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 122280500
Price (USD)/ Each
142.65
Online exclusive
158.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
142.65
Online exclusive
158.00 
Save 15.35 (10%)
Add to cart
Iodoacetic acid, 97%, Thermo Scientific Chemicals
Catalog number122280500
Price (USD)/ Each
142.65
Online exclusive
158.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS64-69-7
IUPAC Name2-iodoacetic acid
Molecular FormulaC2H3IO2
InChI KeyJDNTWHVOXJZDSN-UHFFFAOYSA-N
SMILESOC(=O)CI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange-brown
Appearance (Form)Powder or crystals or flakes
Melting point76°C to 83°C
Infrared spectrumConforms
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only