Thermo Scientific Chemicals

4-Iodoanisole, 98%, Thermo Scientific Chemicals

Catalog number: 122321000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Iodoanisole, 98%, Thermo Scientific Chemicals

Catalog number: 122321000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 122321000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
696-62-8
IUPAC Name
1-iodo-4-methoxybenzene
Molecular Formula
C7H7IO
InChI Key
SYSZENVIJHPFNL-UHFFFAOYSA-N
SMILES
COC1=CC=C(I)C=C1
Appearance (Color)
Off-white to brown
Appearance (Form)
Crystalline powder and/or chunks
Appearance
May darken on storage
Infrared spectrum
Conforms
Melting point
46°C to 51°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text