4-Iodoanisole, 98%
4-Iodoanisole, 98%
4-Iodoanisole, 98%
Thermo Scientific Chemicals

4-Iodoanisole, 98%

CAS: 696-62-8 | C7H7IO | 234.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 122325000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS696-62-8
IUPAC Name1-iodo-4-methoxybenzene
Molecular FormulaC7H7IO
InChI KeySYSZENVIJHPFNL-UHFFFAOYSA-N
SMILESCOC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to brown
Appearance (Form)Crystalline powder and/or chunks
AppearanceMay darken on storage
Infrared spectrumConforms
Melting point46°C to 51°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link