2-Iodobenzoic acid, 98%, Thermo Scientific Chemicals
2-Iodobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Iodobenzoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number 122345000
Price (USD)/ Each
212.65
Online exclusive
236.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
212.65
Online exclusive
236.00 
Save 23.35 (10%)
Add to cart
2-Iodobenzoic acid, 98%, Thermo Scientific Chemicals
Catalog number122345000
Price (USD)/ Each
212.65
Online exclusive
236.00 
Save 23.35 (10%)
-
Add to cart
Chemical Identifiers
CAS88-67-5
IUPAC Name2-iodobenzoic acid
Molecular FormulaC7H5IO2
InChI KeyCJNZAXGUTKBIHP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white to beige
Appearance (Form)Powder
Infrared spectrumConforms
Melting point160°C to 164°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only