Informational:Informational:This product ships via ground from US northeast.
Thermo Scientific Chemicals

(+)-5-Iodo-2'-deoxyuridine, 99%, Thermo Scientific Chemicals

Catalog number: 122350010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

(+)-5-Iodo-2'-deoxyuridine, 99%, Thermo Scientific Chemicals

Catalog number: 122350010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 122350010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
54-42-2
IUPAC Name
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Formula
C9H11IN2O5
InChI Key
XQFRJNBWHJMXHO-RRKCRQDMSA-N
SMILES
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O
Loss on drying
=<1 % (1 g, 60°C) (vacuum)
Specific optical rotation
on dried substance
Titration with NaOH
>=98.5 % (On dried substance)
Appearance (Color)
White to light beige
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text