Thermo Scientific Chemicals

Iodomethane, 99%, stabilized, Thermo Scientific Chemicals

Catalog number: 122370025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Iodomethane, 99%, stabilized, Thermo Scientific Chemicals

Catalog number: 122370025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 122370025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
74-88-4
IUPAC Name
iodomethane
Molecular Formula
CH3I
InChI Key
INQOMBQAUSQDDS-UHFFFAOYSA-N
SMILES
CI
GC
>=98.5 %
Non-volatile matter
=<0.01 %
Infrared spectrum
Conforms
Refractive index
1.5290 to 1.5310 (20°C, 589 nm)
Color scale
=<150 APHA

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text