1-Iodo-4-nitrobenzene, 99%
1-Iodo-4-nitrobenzene, 99%
1-Iodo-4-nitrobenzene, 99%
1-Iodo-4-nitrobenzene, 99%
Thermo Scientific Chemicals

1-Iodo-4-nitrobenzene, 99%

CAS: 636-98-6 | C6H4INO2 | 249.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 122381000
Price (EUR)
181,90
Online Exclusive
214,00
Save 32,10 (15%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
181,90
Online Exclusive
214,00
Save 32,10 (15%)
Each
Add to cart
Chemical Identifiers
IUPAC Name1-iodo-4-nitrobenzene
Molecular FormulaC6H4INO2
InChI KeySCCCFNJTCDSLCY-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(I)C=C1
Molecular Weight (g/mol)249.01
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to orange to brown
Infrared spectrumConforms
GC>=98.5 %
Melting point172°C to 177°C
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link