Thermo Scientific Chemicals

4-Iodophenol, 99%, Thermo Scientific Chemicals

Catalog number: 122390100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Iodophenol, 99%, Thermo Scientific Chemicals

Catalog number: 122390100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 122390100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
540-38-5
IUPAC Name
4-iodophenol
Molecular Formula
C6H5IO
InChI Key
VSMDINRNYYEDRN-UHFFFAOYSA-N
SMILES
OC1=CC=C(I)C=C1
Appearance (Color)
Beige to pink to brown
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Melting point
89°C to 94°C
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text