Chemical Identifiers
CAS146-68-9
IUPAC Name3-(4-iodophenyl)-2-(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular FormulaC19H13ClIN5O2
InChI KeyJORABGDXCIBAFL-UHFFFAOYSA-M
SMILES[Cl-].[O-][N+](=O)C1=CC=C(C=C1)[N+]1=NC(=NN1C1=CC=C(I)C=C1)C1=CC=CC=C1
View more