Isobutyraldehyde, 99+%, Thermo Scientific Chemicals
Isobutyraldehyde, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isobutyraldehyde, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 122510010
Price (USD)/ Each
48.65
Online exclusive
56.60 
Save 7.95 (14%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
48.65
Online exclusive
56.60 
Save 7.95 (14%)
Add to cart
Isobutyraldehyde, 99+%, Thermo Scientific Chemicals
Catalog number122510010
Price (USD)/ Each
48.65
Online exclusive
56.60 
Save 7.95 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-methylpropanal
Molecular FormulaC4H8O
InChI KeyAMIMRNSIRUDHCM-UHFFFAOYSA-N
SMILESCC(C)C=O
Molecular Weight (g/mol)72.11
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3710 to 1.3745 (20°C, 589 nm)
Color scale=<10 APHA
Infrared spectrumConforms
GC>=99.0 %
Free acid=<1.0 % (Isobutyric acid)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only