Thermo Scientific Chemicals

Isobutyraldehyde, 99+%, Thermo Scientific Chemicals

Catalog number: 122510100
10 L, Plastic drum, Each
Thermo Scientific Chemicals

Isobutyraldehyde, 99+%, Thermo Scientific Chemicals

Catalog number: 122510100
10 L, Plastic drum, Each
Quantity
Packaging
Catalog number: 122510100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
78-84-2
IUPAC Name
2-methylpropanal
Molecular Formula
C4H8O
InChI Key
AMIMRNSIRUDHCM-UHFFFAOYSA-N
SMILES
CC(C)C=O
Refractive index
1.3710 to 1.3745 (20°C, 589 nm)
Color scale
=<10 APHA
Infrared spectrum
Conforms
GC
>=99.0 %
Free acid
=<1.0 % (Isobutyric acid)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text