Isobutyric Acid, 99+%
Isobutyric Acid, 99+%
Isobutyric Acid, 99+%
Isobutyric Acid, 99+%
Thermo Scientific Chemicals

Isobutyric Acid, 99+%

CAS: 79-31-2 | C4H8O2 | 88.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number 122520250
Price (USD)
47.65
Online Exclusive
52.30
Save 4.65 (9%)
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
47.65
Online Exclusive
52.30
Save 4.65 (9%)
Each
Add to cart
Chemical Identifiers
CAS79-31-2
IUPAC Name2-methylpropanoic acid
Molecular FormulaC4H8O2
InChI KeyKQNPFQTWMSNSAP-UHFFFAOYSA-N
SMILESCC(C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
Refractive index1.3920 to 1.3940 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link