HR-ICP-MS System Injectors
Product Image
Thermo Scientific™
HR-ICP-MS System Injectors
Meet your HR-ICP-MS analytical requirements with this range of reliable, easy to install Injectors, which ensure that your system is tailored precisely.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
1225270System Injector Kit
1091260Sapphire Iinjector, 1.75 mm
1092000Platinum Injector, 1.8 mm
1122800Quartz injector, 1.75 mm
1130680Quartz injector, 1.0 mm
Catalog number 1225270
Price (USD)
-
Request A Quote
Type:
System Injector Kit
Ask our AI about this Product
Tailor your consumables to your chosen sample matrices for enhanced analysis with this range of Thermo Scientific™ HR-ICP-MS System Injectors for the Thermo Scientific™ ELEMENT™ Series HR-ICP-MS system.
This wide selection of reliable, easy to install Injectors, with differing internal diameters and materials, ensures that your system is optimized precisely to meet your analytical requirements. All our injectors are easily removable for improved robustness and reduced maintenance requirements.
Choose High Quality Consumables to Improve Performance
  • Quartz injector (1.75mm I.D.) for standard torch
  • Quartz injector (1.0mm I.D.) for standard torch, for organic matrices
  • Sapphire injector (1.75mm I.D.) for standard torch, for HF-containing solutions
  • Platinum injector (1.8mm I.D.) for standard torch
  • Platinum injector (1.0mm I.D.) for standard torch, for organic matrices
  • Dual Mode Interface (dual mode torch holder assembly, dual mode torch injector, cyclone dual mode spray chamber, nebulizer) for GC-coupling
Specifications
For Use With (Equipment)ELEMENT Series HR-ICP-MS System
TypeSystem Injector Kit
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link