Thermo Scientific Chemicals

Isochroman, 99%, Thermo Scientific Chemicals

Catalog number: 122540050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Isochroman, 99%, Thermo Scientific Chemicals

Catalog number: 122540050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 122540050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
493-05-0
IUPAC Name
3,4-dihydro-1H-2-benzopyran
Molecular Formula
C9H10O
InChI Key
HEBMCVBCEDMUOF-UHFFFAOYSA-N
SMILES
C1CC2=CC=CC=C2CO1
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.5430 to 1.5460 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text