Isoeugenol, 98+%, mixture of cis/trans isomers, Thermo Scientific Chemicals
Isoeugenol, 98+%, mixture of cis/trans isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isoeugenol, 98+%, mixture of cis/trans isomers, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
Glass bottle
Catalog number 122571000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS97-54-1
IUPAC Name2-methoxy-4-[(1E)-prop-1-en-1-yl]phenol
Molecular FormulaC10H12O2
InChI KeyBJIOGJUNALELMI-ONEGZZNKSA-N
SMILESCOC1=CC(\C=C\C)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Viscous liquid
Infrared spectrumConforms
GC>=98.0 %
Refractive index1.5750 to 1.5790 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only