Thermo Scientific Chemicals

Isonicotinic acid, 99%, Thermo Scientific Chemicals

Catalog number: 122595000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Isonicotinic acid, 99%, Thermo Scientific Chemicals

Catalog number: 122595000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 122595000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
55-22-1
IUPAC Name
pyridine-4-carboxylic acid
Molecular Formula
C6H5NO2
InChI Key
TWBYWOBDOCUKOW-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=NC=C1
Infrared spectrum
Conforms
Appearance (Color)
White to cream
Titration with NaOH
>=98.5 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text