The PureLink® Foil Tape is a component in the PureLink® 96 Plasmid Purification System. The PureLink® 96 Plasmid Purification System offers a unique approach to high throughput plasmid DNA isolation.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Color
Silver
For Use With (Application)
Automated Nucleic Acid Purification, DNA Extraction
Plate Compatibility
384-well Plates, 96-well Plates
Product Line
PureLink™
Product Type
Foil Tape
Quantity
50 Pieces
Shipping Condition
Room Temperature
Contents & Storage
Package of 50 pieces The PureLink™ 96 Components are shipped at room temperature. Upon receipt, store the components at room temperature.