Thermo Scientific Chemicals

Isonipecotamide, 98%, Thermo Scientific Chemicals

Catalog number: 122615000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Isonipecotamide, 98%, Thermo Scientific Chemicals

Catalog number: 122615000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 122615000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
39546-32-2
IUPAC Name
piperidine-4-carboxamide
Molecular Formula
C6H12N2O
InChI Key
DPBWFNDFMCCGGJ-UHFFFAOYSA-N
SMILES
NC(=O)C1CCNCC1
Appearance (Form)
Crystalline powder
Melting point
146°C to 150°C (on dry substance)
Loss on drying
=<2 % (70°C)
Appearance (Color)
White to light yellow
Water
=<2 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text