Isonipecotic Acid, 99%
Isonipecotic Acid, 99%
Thermo Scientific Chemicals

Isonipecotic Acid, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 122620250
Price (USD)/ Each
54.10
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
54.10
Add to cart
Isonipecotic Acid, 99%
Catalog number122620250
Price (USD)/ Each
54.10
-
Add to cart
Chemical Identifiers
CAS498-94-2
IUPAC Namepiperidine-4-carboxylic acid
Molecular FormulaC6H11NO2
InChI KeySRJOCJYGOFTFLH-UHFFFAOYSA-N
SMILESOC(=O)C1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pink-beige
Appearance (Form)Powder
Solubility(10 % in water) Clear to slightly hazy
Infrared spectrumConforms
Titration with HClO4>=98.5 % (On dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only