Isophorone, 97%, Thermo Scientific Chemicals
Isophorone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isophorone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 122640010
Price (USD)/ Each
35.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
41.70 
Save 6.05 (15%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
35.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
41.70 
Save 6.05 (15%)
Add to cart
Isophorone, 97%, Thermo Scientific Chemicals
Catalog number122640010
Price (USD)/ Each
35.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
41.70 
Save 6.05 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name3,5,5-trimethylcyclohex-2-en-1-one
Molecular FormulaC9H14O
InChI KeyHJOVHMDZYOCNQW-UHFFFAOYSA-N
SMILESCC1=CC(=O)CC(C)(C)C1
Molecular Weight (g/mol)138.21
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4750 to 1.4800 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only