Isophorone, 97%
Isophorone, 97%
Isophorone, 97%
Thermo Scientific Chemicals

Isophorone, 97%

CAS: 78-59-1 | C9H14O | 138.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 122645000
Price (USD)
22.65
Special offer
Online exclusive
Ends: 26-Sep-2025
28.20
Save 5.55 (20%)
Each
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
22.65
Special offer
Online exclusive
Ends: 26-Sep-2025
28.20
Save 5.55 (20%)
Each
Add to cart
Chemical Identifiers
CAS78-59-1
IUPAC Name3,5,5-trimethylcyclohex-2-en-1-one
Molecular FormulaC9H14O
InChI KeyHJOVHMDZYOCNQW-UHFFFAOYSA-N
SMILESCC1=CC(=O)CC(C)(C)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4750 to 1.4800 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link