Thermo Scientific Chemicals

Isophthalic acid, 99%, Thermo Scientific Chemicals

Catalog number: 122650020
2 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Isophthalic acid, 99%, Thermo Scientific Chemicals

Catalog number: 122650020
2 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 122650020
Price (USD)
Have Questions?

Chemical Identifiers

CAS
121-91-5
IUPAC Name
benzene-1,3-dicarboxylic acid
Molecular Formula
C8H6O4
InChI Key
QQVIHTHCMHWDBS-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(=CC=C1)C(O)=O
Titration with NaOH
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text