Thermo Scientific Chemicals

Isophthaloyl dichloride, 98%, Thermo Scientific Chemicals

Catalog number: 122661000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Isophthaloyl dichloride, 98%, Thermo Scientific Chemicals

Catalog number: 122661000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 122661000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-63-8
IUPAC Name
benzene-1,3-dicarbonyl dichloride
Molecular Formula
C8H4Cl2O2
InChI Key
FDQSRULYDNDXQB-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC(=CC=C1)C(Cl)=O
Appearance (Color)
White
Appearance (Form)
Crystalline mass
Infrared spectrum
Conforms
Melting point
43°C to 45°C
Titration RCOCl
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text