Isopropyl disulfide, 96%, Thermo Scientific Chemicals
Isopropyl disulfide, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isopropyl disulfide, 96%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
25 g
Packaging:
Glass bottle
Catalog number 122700250
Price (USD)/ Each
43.30
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
43.30
Add to cart
Isopropyl disulfide, 96%, Thermo Scientific Chemicals
Catalog number122700250
Price (USD)/ Each
43.30
-
Add to cart
Chemical Identifiers
CAS4253-89-8
IUPAC Name2-(propan-2-yldisulfanyl)propane
Molecular FormulaC6H14S2
InChI KeyLZAZXBXPKRULLB-UHFFFAOYSA-N
SMILESCC(C)SSC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4890 to 1.4930 (20°C, 589 nm)
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only