Chemical Identifiers
CAS362-75-4
IUPAC Name[(3aR,4R,6R,6aR)-6-(6-amino-9H-purin-9-yl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl]methanol
Molecular FormulaC13H17N5O4
InChI KeyLCCLUOXEZAHUNS-AUWRGFAENA-N
SMILESCC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)N1C=NC2=C(N)N=CN=C12
View more