Thermo Scientific Chemicals

2',3'-O-Isopropylideneadenosine, 98%

Catalog number: 122720250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2',3'-O-Isopropylideneadenosine, 98%

Catalog number: 122720250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 122720250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
362-75-4
IUPAC Name
[(3aR,4R,6R,6aR)-6-(6-amino-9H-purin-9-yl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl]methanol
Molecular Formula
C13H17N5O4
InChI Key
LCCLUOXEZAHUNS-AUWRGFAENA-N
SMILES
CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)N1C=NC2=C(N)N=CN=C12
UV spectrophotometry
>=96 % (lambda max. 259 nm, pH 7)
UV spectrophotometry
molar absorptivity 15400 +/-4%, on dry substance
Ratio of Absorbances
0.75 to 0.81 (A250/A260)
Ratio of Absorbances
0.13 to 0.17 (A280/A260)
Ratio of Absorbances
=<0.01 (A290/A260)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text