Thermo Scientific Chemicals

Itaconic acid, 99+%, Thermo Scientific Chemicals

Catalog number: 122810010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Itaconic acid, 99+%, Thermo Scientific Chemicals

Catalog number: 122810010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 122810010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
97-65-4
IUPAC Name
2-methylidenebutanedioic acid
Molecular Formula
C5H6O4
InChI Key
LVHBHZANLOWSRM-UHFFFAOYSA-N
SMILES
OC(=O)CC(=C)C(O)=O
Melting point
165°C to 168°C
Titration with NaOH
>=99.0 %
Heavy metals (as Pb)
=<20 ppm
Appearance (Form)
Crystalline powder or crystals
Sulfate (SO4)
=<300 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text