Itaconic acid, 99+%, Thermo Scientific Chemicals
Itaconic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Itaconic acid, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number 122810051
Price (USD)/ Each
159.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
188.00 
Save 28.35 (15%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
159.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
188.00 
Save 28.35 (15%)
Add to cart
Itaconic acid, 99+%, Thermo Scientific Chemicals
Catalog number122810051
Price (USD)/ Each
159.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
188.00 
Save 28.35 (15%)
-
Add to cart
Chemical Identifiers
CAS97-65-4
IUPAC Name2-methylidenebutanedioic acid
Molecular FormulaC5H6O4
InChI KeyLVHBHZANLOWSRM-UHFFFAOYSA-N
SMILESOC(=O)CC(=C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point165°C to 168°C
Titration with NaOH>=99.0 %
Heavy metals (as Pb)=<20 ppm
Appearance (Form)Crystalline powder or crystals
Sulfate (SO4)=<300 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only