Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

PureLink™ Viral Lysis Buffer

Catalog number: 12282500
Have Questions?
Invitrogen™

PureLink™ Viral Lysis Buffer

Catalog number: 12282500
Have Questions?
Catalog Number
12282500
also known as 12282-500
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
12282500
also known as 12282-500
500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The PureLink™ 96 Viral RNA/DNA Kit is a 96-well silica plate system that can extract both viral RNA and DNA from 200-μl cell-free fluids which uses a low-depth 96-well centrifuge bucket for ease-of- use, all without any loss in sensitivity. These kits provide flexibility in centrifuge choice. Minimum speeds for 96 well processing of 2250 x g and a rotor bucket depth of 5 cm make this kit one of the most adaptable systems for existing laboratory centrifuges
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer
Lysis Buffer
For Use With (Application)
Nucleic Acid Extraction
Product Line
PureLink™
Product Type
Viral Lysis Buffer
Quantity
500 mL
Shipping Condition
Room Temperature

Contents & Storage

The PureLink™ 96 Viral RNA/DNA Kit contains a PureLink™ Filter Plate, Receiver plates, deep-well blocks, foil tape, carrier RNA, proteinase K, Viral Lysis Buffer, Wash Buffer, and RNase-free Water. The kit is shipped and stored at room temperature and the lyophilized tRNA should be removed and stored at -20°C upon receipt. The proteinase K should be stored at +4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text