DynaMag™-50 Magnet
DynaMag™-50 Magnet
Invitrogen™

DynaMag™-50 Magnet

The new DynaMag™-50 magnet is optimized for efficient cell isolation using Dynabeads™. The DynaMag™-50 magnet combines a strong magnetic attractionRead more
Have Questions?
Catalog NumberQuantity
12302D1 Each
Catalog number 12302D
Price (USD)
1,232.65
Online Exclusive
1,354.00
Save 121.35 (9%)
Each
In stock
Add to cart
Quantity:
1 Each
Price (USD)
1,232.65
Online Exclusive
1,354.00
Save 121.35 (9%)
Each
Add to cart
The new DynaMag™-50 magnet is optimized for efficient cell isolation using Dynabeads™. The DynaMag™-50 magnet combines a strong magnetic attraction for efficient separations with a flexible and smart ergonomic design that ensures a comfortable working position. A tilting mechanism on the magnet enables you to rest your elbows while pipetting. The magnet can be used for any starting sample in combination with all types of cell-isolation Dynabeads™.
 
→ Optimal working volume: 5–50 ml
→ Magnet holds 2 x 50 ml tubes
→ Excellent control and visibility of your isolations
 
 

NOTE: This magnet replaces the Dynal MPC™-50 magnet
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)DynaMag™
Quantity1 Each
Sample Volume10-50 mL
Shipping ConditionRoom Temperature
Product LineDynaMag™
TypeCell Separation Magnet
Unit SizeEach
Contents & Storage
Store at room temperature.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection